CAS RN 84209-07-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-(N-phthalimidooxy)-2-methylpropionate, 98%, Thermo Scientific™
CAS: 84209-07-4 Synonym: Methyl 2-(1,3-dioxo-1,3-dihydro-2-isoindolyloxy)-2-methylpropionate