CAS RN 84249-39-8
IUPAC Name:
5-nitro-N-(1-phenylethyl)pyridin-2-amine
Synonyms:
5-nitro-N-(1-phenylethyl)pyridin-2-amine
Molecular Weight (g/mol):
243.27
Molecular Formula:
C13H13N3O2
InChi Key:
RAKMWGKBCDCUDX-UHFFFAOYNA-N
SMILES:
CC(NC1=NC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1