accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSN

CAS RN 84302-57-8

IUPAC Name: 2-methyl-4-phenyl-1,3-thiazole-5-carbonitrile
Synonyms: 2-methyl-4-phenyl-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol): 200.26
Molecular Formula: C11H8N2S
InChi Key: CLGMFVPFRMXIRG-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results