accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClOH

CAS RN 84367-31-7

IUPAC Name: 4-chloro-3-hydroxybutanenitrile
Synonyms: 4-chloro-3-hydroxybutanenitrile
Molecular Weight (g/mol): 119.55
Molecular Formula: C4H6ClNO
InChi Key: LHBPNZDUNCZWFL-UHFFFAOYNA-N
SMILES: OC(CCl)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results