accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2ClONHNOHOOHO(Z)

CAS RN 84370-95-6

IUPAC Name: (2Z)-but-2-enedioic acid; 4-amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide
Synonyms: clebopride; maleic acid
Molecular Weight (g/mol): 489.95
Molecular Formula: C24H28ClN3O6
InChi Key: BCVIWCRZYPHHMQ-BTJKTKAUSA-N
SMILES: OC(=O)\C=C/C(O)=O.COC1=CC(N)=C(Cl)C=C1C(=O)NC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results