Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Clebopride maleate

Catalog No. AC465412500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465412500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465412500 Supplier Thermo Scientific Chemicals Supplier No. 465412500
 Restricted Item
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84370-95-6
Molecular Formula C24H28ClN3O6
Molecular Weight (g/mol) 489.95
InChI Key BCVIWCRZYPHHMQ-BTJKTKAUSA-N
IUPAC Name (2Z)-but-2-enedioic acid; 4-amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide
SMILES OC(=O)\C=C/C(O)=O.COC1=CC(N)=C(Cl)C=C1C(=O)NC1CCN(CC2=CC=CC=C2)CC1

Specifications

Quantity 250 mg
Formula Weight 489.95
Chemical Name or Material Clebopride maleate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.