accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84476-99-3

IUPAC Name: 2,5-difluoropyridine
Synonyms: 2,5-difluoropyridine
Molecular Weight (g/mol): 115.08
Molecular Formula: C5H3F2N
InChi Key: XFAMUOYNXFXQTC-UHFFFAOYSA-N
SMILES: FC1=CN=C(F)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results