Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoropyridine 98.0+%, TCI America™
SDP

Catalog No. D41745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41745G 5 g
D41741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41745G Supplier TCI America Supplier No. D41745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84476-99-3
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD04114191
InChI Key XFAMUOYNXFXQTC-UHFFFAOYSA-N
PubChem CID 2762871
IUPAC Name 2,5-difluoropyridine
SMILES C1=CC(=NC=C1F)F

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 115°C
Quantity 5 g
UN Number 1993
Synonym pyridine, 2,5-difluoro, 2,5-difluorpyridin, pubchem14398, acmc-209put, pyridine,2,5-difluoro, 2,5-bis fluoranyl pyridine, ksc652g9r, 2,5-difluoropyridine, abbypharma ap-14-5383
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.