accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFClNOO

CAS RN 84478-75-1

IUPAC Name: 2-chloro-4-fluoro-5-nitrophenol
Synonyms: 2-chloro-4-fluoro-5-nitrophenol
Molecular Weight (g/mol): 191.54
Molecular Formula: C6H3ClFNO3
InChi Key: NAWVMCKMQMJQMF-UHFFFAOYSA-N
SMILES: OC1=CC(=C(F)C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results