CAS RN 84484-78-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BTS 54-505 hydrochloride
Potent SNRI; active metabolite of sibutramine