accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrH2NBr

CAS RN 84539-34-4

IUPAC Name: 3,5-dibromopyridin-4-amine
Synonyms: 3,5-dibromopyridin-4-amine
Molecular Weight (g/mol): 251.91
Molecular Formula: C5H4Br2N2
InChi Key: MNRBVTDLMUNVLD-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=NC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results