accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHHCl(?)

CAS RN 84540-61-4

IUPAC Name: N-(1-methylpiperidin-4-ylidene)hydroxylamine hydrochloride
Synonyms: N-(1-methylpiperidin-4-ylidene)hydroxylamine hydrochloride
Molecular Weight (g/mol): 164.63
Molecular Formula: C6H13ClN2O
InChi Key: WQVZFAGWTNLHSX-UHFFFAOYSA-N
SMILES: Cl.CN1CCC(CC1)=NO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results