CAS RN 845533-86-0
IUPAC Name:
1-(6-bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol; but-2-enedioic acid
Synonyms:
bedaquiline; butenedioic acid
Molecular Weight (g/mol):
671.59
Molecular Formula:
C36H35BrN2O6
InChi Key:
ZLVSPMRFRHMMOY-UHFFFAOYNA-N
SMILES:
OC(=O)C=CC(O)=O.COC1=C(C=C2C=C(Br)C=CC2=N1)C(C1=CC=CC=C1)C(O)(CCN(C)C)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Bedaquiline fumarate
Thermo Scientific Chemicals Bedaquiline fumarate
CAS: 845533-86-0 Molecular Formula: C32H31BrN2O2·C4H4O4