CAS RN 84563-54-2
IUPAC Name:
bis(4-tert-butylphenyl)iodanium trifluoromethanesulfonate
Synonyms:
bis(4-tert-butylphenyl)iodanium triflate
Molecular Weight (g/mol):
542.4
Molecular Formula:
C21H26F3IO3S
InChi Key:
VGZKCAUAQHHGDK-UHFFFAOYSA-M
SMILES:
[O-]S(=O)(=O)C(F)(F)F.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Bis(4-tert-butylphenyl)iodonium triflate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.