accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOHH2NHCl

CAS RN 845861-49-6

IUPAC Name: 3-(2-aminoethyl)-2-methyl-1H-indol-5-ol hydrochloride
Synonyms: 2-methyl-5-HT hydrochloride
Molecular Weight (g/mol): 226.7
Molecular Formula: C11H15ClN2O
InChi Key: ZNVRHQPXSBWLQP-UHFFFAOYSA-N
SMILES: Cl.CC1=C(CCN)C2=CC(O)=CC=C2N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results