accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COBON

CAS RN 845885-86-1

IUPAC Name: 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
Synonyms: 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
Molecular Weight (g/mol): 191.04
Molecular Formula: C10H14BNO2
InChi Key: HDQULWCYQQMIJJ-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CC=NC=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results