accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84678-52-4

IUPAC Name: 9-ethyl-3-[(2-ethyl-2-phenylhydrazin-1-ylidene)methyl]-9H-carbazole
Synonyms: 9-ethyl-3-[(2-ethyl-2-phenylhydrazin-1-ylidene)methyl]carbazole
Molecular Weight (g/mol): 341.46
Molecular Formula: C23H23N3
InChi Key: IPCPWEQNRXADBE-UHFFFAOYSA-N
SMILES: CCN(N=CC1=CC2=C(C=C1)N(CC)C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results