Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Ethylcarbazole-3-carboxaldehyde N-Ethyl-N-phenylhydrazone 98.0+%, TCI America™
SDP

Catalog No. E05761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05761G Supplier TCI America Supplier No. E05761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84678-52-4
Molecular Formula C23H23N3
Molecular Weight (g/mol) 341.458
MDL Number MFCD03208617
InChI Key IPCPWEQNRXADBE-JJIBRWJFSA-N
Synonym 9-Ethyl-3-(N-ethyl-N-phenylhydrazonomethyl)carbazole
PubChem CID 9608395
IUPAC Name N-ethyl-N-[(E)-(9-ethylcarbazol-3-yl)methylideneamino]aniline
SMILES CCN1C2=C(C=C(C=C2)C=NN(CC)C3=CC=CC=C3)C4=CC=CC=C41

Specifications

Color Yellow
Quantity 1 g
Formula Weight 341.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Ethylcarbazole-3-carboxaldehyde N-Ethyl-N-phenylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.