accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84682-27-9

IUPAC Name: 1-(4-methylphenyl)cyclohexane-1-carboxylic acid
Synonyms: 1-(4-methylphenyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 218.3
Molecular Formula: C14H18O2
InChi Key: BEVYUARQHKRBMH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1(CCCCC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results