Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(p-Tolyl)-1-cyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. T30975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30975G 5 g
T309725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30975G Supplier TCI America Supplier No. T30975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84682-27-9
Molecular Formula C14H18O2
Molecular Weight (g/mol) 218.30
MDL Number MFCD00065264
InChI Key BEVYUARQHKRBMH-UHFFFAOYSA-N
Synonym 1-4-methylphenyl cyclohexanecarboxylic acid, 1-4-methylphenyl-1-cyclohexanecarboxylic acid, 1-p-tolyl cyclohexanecarboxylic acid, 1-p-tolyl-cyclohexanecarboxylic acid, 1-4-methylphenyl cyclohexane-1-carboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylic acid, 1-p-tolylcyclohexanecarboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylicacid, cyclohexanecarboxylicacid, 1-4-methylphenyl, cyclohexanecarboxylic acid, 1-4-methylphenyl
PubChem CID 98664
IUPAC Name 1-(4-methylphenyl)cyclohexane-1-carboxylic acid
SMILES CC1=CC=C(C=C1)C1(CCCCC1)C(O)=O

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(p-Tolyl)-1-cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.