CAS RN 84687-43-4
CAS RN 84687-43-4
IUPAC Name:
(2R,3R,4S,5S,6R)-2-{[(3R,9R,12S,14R,15R,16R)-14-hydroxy-15-[(2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyloxolan-2-yl]-7,7,12,16-tetramethyl-6-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-9-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
(2R,3R,4S,5S,6R)-2-{[(3R,9R,12S,14R,15R,16R)-14-hydroxy-15-[(2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyloxolan-2-yl]-7,7,12,16-tetramethyl-6-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-9-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
784.98
Molecular Formula:
C41H68O14
InChi Key:
QMNWISYXSJWHRY-CSXKERSZSA-N
SMILES:
CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]1[C@H](O)C[C@@]2(C)C3C[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C4[C@]5(CC35CC[C@]12C)CCC(O[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O)C4(C)C
1
–
1
of
1
results
1
Astragaloside IV, Thermo Scientific™
CAS: 84687-43-4 Molecular Formula: C41 H68 O14