accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHHOH3CH3CCH3CH3(E/Z)

CAS RN 847239-17-2

IUPAC Name: 3-{4-hydroxy-3-[5,5,8,8-tetramethyl-3-(pentyloxy)-5,6,7,8-tetrahydronaphthalen-2-yl]phenyl}prop-2-enoic acid
Synonyms: 3-{4-hydroxy-3-[5,5,8,8-tetramethyl-3-(pentyloxy)-6,7-dihydronaphthalen-2-yl]phenyl}prop-2-enoic acid
Molecular Weight (g/mol): 436.59
Molecular Formula: C28H36O4
InChi Key: APJSHECCIRQQDV-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC2=C(C=C1C1=C(O)C=CC(C=CC(O)=O)=C1)C(C)(C)CCC2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results