accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84752-61-4

NHSOOONOO

CAS RN 84752-61-4

IUPAC Name: pyridin-1-ium 3-nitrobenzene-1-sulfonate
Synonyms: pyridium 3-nitrobenzenesulfonate
Molecular Weight (g/mol): 282.27
Molecular Formula: C11H10N2O5S
InChi Key: WZZFZXZRKPNZOC-UHFFFAOYSA-N
SMILES: C1=CC=[NH+]C=C1.[O-][N+](=O)C1=CC(=CC=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results