accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOOONi(E/Z)(E/Z)

CAS RN 847654-17-5

IUPAC Name: nickel(2+) 2-{[(2-{[1-oxido-2-(piperidin-1-yl)ethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate
Synonyms: nickel(2+) 2-{[(2-{[1-oxido-2-(piperidin-1-yl)ethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate
Molecular Weight (g/mol): 436.14
Molecular Formula: C22H23N3NiO3
InChi Key: WUBYDZLSGCSGEK-UHFFFAOYSA-L
SMILES: [Ni++].[O-]C(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1N=C([O-])CN1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results