accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3OHSOOFNON

CAS RN 84806-27-9

IUPAC Name: 7-fluoro-2,1,3-benzoxadiazole-4-sulfonic acid amine
Synonyms: 7-fluoro-2,1,3-benzoxadiazole-4-sulfonic acid amine
Molecular Weight (g/mol): 235.19
Molecular Formula: C6H6FN3O4S
InChi Key: JXLHNMVSKXFWAO-UHFFFAOYSA-N
SMILES: N.OS(=O)(=O)C1=CC=C(F)C2=NON=C12
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)