Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluorobenzo-2-oxa-1,3-diazole-4-sulfonic acid ammonium salt, MP Biomedicals™

Catalog No. ICN15808625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15808625 25 mg
ICN15808605 5 mg
ICN15808610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15808625 Supplier MP Biomedicals, Inc Supplier No. 0215808625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sensitive and specific fluorescent labeling reagent for low molecular weight compounds and for macromolecules containing thiol groups

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84806-27-9
Molecular Formula C6H6FN3O4S
Molecular Weight (g/mol) 235.189
MDL Number MFCD02682109
InChI Key JXLHNMVSKXFWAO-UHFFFAOYSA-N
Synonym sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
PubChem CID 2737780
IUPAC Name azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
SMILES C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Specifications

Quantity 25 mg
Formula Weight 235.2
Physical Form Powder
Chemical Name or Material 7-Fluorobenzo-2-oxa-1, 3-diazole-4-sulfonic acid ammonium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.