accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 848133-76-6

IUPAC Name: N-(4-chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
Synonyms: N-(4-chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
Molecular Weight (g/mol): 289.72
Molecular Formula: C14H12ClN3O2
InChi Key: XDXGFTCQRAQEEG-UHFFFAOYSA-N
SMILES: CCOC1=C(NC(C)=O)C=C2C(Cl)=C(C=NC2=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results