accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OH

CAS RN 84852-15-3

IUPAC Name: 4-(7-methyloctyl)phenol
Synonyms: 4-(7-methyloctyl)phenol
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: JSFITYFUKSFPBZ-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)