Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nonylphenol, 99%, mixture of isomers

Catalog No. AC416240010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416240010 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC416240010 Supplier Thermo Scientific Chemicals Supplier No. 416240010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84852-15-3 | C15H24O | 220.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84852-15-3
MDL Number MFCD00002396
InChI Key SNQQPOLDUKLAAF-UHFFFAOYSA-N
Synonym phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
PubChem CID 67296
IUPAC Name 2-nonylphenol
SMILES CCCCCCCCCC1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.9400g/mL
Boiling Point 293.0°C to 297.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C15H24O
Refractive Index 1.5105 to 1.5165
Quantity 1 kg
Specific Gravity 0.94
Merck Index 14, 6681
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in diluted aq. naoh,soluble in benzene,chlorinated solvents,aniline,,alchols,ethylene glycol
Molecular Weight (g/mol) 220.35
Formula Weight 220.35
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 4-Nonylphenol, Mixture of isomers
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 284-325-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.