accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 848658-70-8

ONOCl

CAS RN 848658-70-8

IUPAC Name: 3-(chloromethyl)-5-(furan-2-yl)-1,2-oxazole
Synonyms: 3-(chloromethyl)-5-(furan-2-yl)-1,2-oxazole
Molecular Weight (g/mol): 183.59
Molecular Formula: C8H6ClNO2
InChi Key: IPIMDVOPXIOKAK-UHFFFAOYSA-N
SMILES: ClCC1=NOC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results