accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH3CCH3NNNNClNH2

CAS RN 848695-25-0

IUPAC Name: 6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]-9H-purin-2-amine
Synonyms: 6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]purin-2-amine
Molecular Weight (g/mol): 318.77
Molecular Formula: C14H15ClN6O
InChi Key: QULDDKSCVCJTPV-UHFFFAOYSA-N
SMILES: COC1=C(C)C(CN2C=NC3=C(Cl)N=C(N)N=C23)=NC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results