accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 848779-87-3

IUPAC Name: [2-(benzyloxy)-4-fluorophenyl]boronic acid
Synonyms: 2-(benzyloxy)-4-fluorophenylboronic acid
Molecular Weight (g/mol): 246.04
Molecular Formula: C13H12BFO3
InChi Key: IFPQNCKVOQKEIU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C=C1OCC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results