accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSOOHNN

CAS RN 849136-14-7

IUPAC Name: 3-bromo-N-[(pyridin-3-yl)methyl]benzene-1-sulfonamide
Synonyms: 3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide
Molecular Weight (g/mol): 327.2
Molecular Formula: C12H11BrN2O2S
InChi Key: VAQOTDUYHINTAB-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)S(=O)(=O)NCC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results