accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 849228-34-8

CH3NHSOOBrCH3

CAS RN 849228-34-8

IUPAC Name: 3-bromo-N-(2,3-dimethylphenyl)benzene-1-sulfonamide
Synonyms: 3-bromo-N-(2,3-dimethylphenyl)benzenesulfonamide
Molecular Weight (g/mol): 340.24
Molecular Formula: C14H14BrNO2S
InChi Key: GAZGMCMRCZAATD-UHFFFAOYSA-N
SMILES: CC1=C(C)C(NS(=O)(=O)C2=CC(Br)=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results