accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84955-31-7

IUPAC Name: 4-chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine
Synonyms: 4-chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine
Molecular Weight (g/mol): 168.58
Molecular Formula: C6H5ClN4
InChi Key: VIVLSUIQHWGALQ-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=C2C=CNC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results