CAS RN 849681-64-7
IUPAC Name:
4,4,5,5-tetramethyl-2-(4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl}phenyl)-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-(4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl}phenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol):
430.16
Molecular Formula:
C26H32B2O4
InChi Key:
QJTLCFKOLMALPJ-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Sigma Aldrich 1-Butyl-1-Methylpyrrolidinium Trifluoromethanesulfonate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.