Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-1-Methylpyrrolidinium Trifluoromethanesulfonate

Catalog No. 72772550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-772-550G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-772-550G Supplier Sigma Aldrich Supplier No. 72772550G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate ([BMPy][OTf]) is an ionic liquid that can be used as a solvent in: Rhodium-catalyzed regioselective hydroformylation reactions. Direct asymmetric aldol condensation reaction. Desulfurization of fuels. Nucleophilic aromatic substitution reactions.([BMPy][OTf]) can also be used as an electrolyte in supercapacitor applications.

Specifications

CAS 849681-64-7
Melting Point 147°C to 152°C
Molecular Formula C4H7BF3K
Linear Formula C4H7BF3K
MDL Number MFCD16621473
Quantity 5 g
Molecular Weight (g/mol) 162 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.