accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 849773-97-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 824.4
Molecular Formula: C38H60BF4FeP2Rh
InChi Key:
SMILES: [Fe].[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results