Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™

Catalog No. AC437292500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437292500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC437292500 Supplier Thermo Scientific Chemicals Supplier No. 437292500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 849773-97-3
Molecular Formula C38H60BF4FeP2Rh
Molecular Weight (g/mol) 824.40
MDL Number MFCD15144872
InChI Key MFSWTFGIDMKBOY-UHFFFAOYNA-N
Synonym 1,1 inverted exclamation marka-bis 2r,5r-2,5-diisopropylphospholano ferrocene cyclooctadiene rhodium i tetrafluoroborate
PubChem CID 71310759
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-cyclopenta-2,4-dien-1-yl-2,5-di(propan-2-yl)phospholane;iron(2+);rhodium;tetrafluoroborate
SMILES [Fe].[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1.CC(C)[C@H]1CC[C@H](C(C)C)P1c1cccc1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 235.0°C to 238.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 824.39
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,1′-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluorobor
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.