accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85-52-9

IUPAC Name: 2-benzoylbenzoic acid
Synonyms: 2-benzoylbenzoic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: FGTYTUFKXYPTML-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)