Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Benzoylbenzoic Acid, MP Biomedicals

Catalog No. ICN213371 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213371 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213371 Supplier MP Biomedicals, Inc Supplier No. 0521337180

Chemical Identifiers

CAS 85-52-9
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002472
InChI Key FGTYTUFKXYPTML-UHFFFAOYSA-N
Synonym o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
PubChem CID 6813
IUPAC Name 2-benzoylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 226.23
Chemical Name or Material o-Benzoylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.