accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNN(E/Z)

CAS RN 85-84-7

IUPAC Name: 1-(2-phenyldiazen-1-yl)naphthalen-2-amine
Synonyms: 1-(2-phenyldiazen-1-yl)naphthalen-2-amine
Molecular Weight (g/mol): 247.3
Molecular Formula: C16H13N3
InChi Key: KLCDQSGLLRINHY-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC=CC2=C1N=NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results