Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yellow AB, MP Biomedicals

Catalog No. ICN204203 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204203 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204203 Supplier MP Biomedicals, Inc Supplier No. 0520420310

Chemical Identifiers

CAS 85-84-7
Molecular Formula C16H13N3
Molecular Weight (g/mol) 247.301
InChI Key KLCDQSGLLRINHY-UHFFFAOYSA-N
PubChem CID 6824
IUPAC Name 1-phenyldiazenylnaphthalen-2-amine
SMILES C1=CC=C(C=C1)N=NC2=C(C=CC3=CC=CC=C32)N

Specifications

Formula Weight 247.3
Quantity 10 g
Chemical Name or Material Yellow Ab
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.