CAS RN 85-85-8
1-(2-Pyridylazo)-2-naphthol, 98%
CAS: 85-85-8 Molecular Formula: C15H11N3O Molecular Weight (g/mol): 249.273 MDL Number: MFCD00004071 InChI Key: RAXUMGMWXZYADR-OBGWFSINSA-N Synonym: 1-2-pyridylazo-2-naphthol,unii-095b53y3xv,pan indicator,2-naphthalenol, 1-2-pyridinylazo,1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one,1-2-pyridylazo-2-hydroxynaphthalene,pan van,1-2-pyridyldiazenyl naphthalen-2-ol,1-pyridin-2-azo-2-naphthol,2-naphthalenol, 1-2-2-pyridinyl diazenyl PubChem CID: 5376264 IUPAC Name: (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one SMILES: C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
Thermo Scientific Chemicals 1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
CAS: 85-85-8 InChI Key: RAXUMGMWXZYADR-OBGWFSINSA-N Synonym: 1-2-pyridylazo-2-naphthol,unii-095b53y3xv,pan indicator,2-naphthalenol, 1-2-pyridinylazo,1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one,1-2-pyridylazo-2-hydroxynaphthalene,pan van,1-2-pyridyldiazenyl naphthalen-2-ol,1-pyridin-2-azo-2-naphthol,2-naphthalenol, 1-2-2-pyridinyl diazenyl PubChem CID: 5376264 IUPAC Name: (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one SMILES: C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
PAN Indicator, 0.1% Solution in Ethanol, Spectrum™ Chemical
CAS: 85-85-8 Molecular Formula: C15H11N3O Molecular Weight (g/mol): 249.27 InChI Key: RAXUMGMWXZYADR-SDXDJHTJSA-N IUPAC Name: (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1
PAN, 1% (w/v) Indicator Solution in Methanol, Spectrum™ Chemical
Analytical indicator solution for laboratory use.
PAN, 0.1% (w/v) Indicator Solution in Methanol, Spectrum™ Chemical
CAS: 85-85-8 Molecular Formula: C15H11N3O Molecular Weight (g/mol): 249.27 InChI Key: RAXUMGMWXZYADR-SDXDJHTJSA-N IUPAC Name: (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1
PAN, Reagent, ACS, Spectrum™ Chemical
CAS: 85-85-8 Molecular Formula: C15H11N3O Molecular Weight (g/mol): 249.27 InChI Key: RAXUMGMWXZYADR-SDXDJHTJSA-N IUPAC Name: (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1
PAN 98.0+%, TCI America™
CAS: 85-85-8 Molecular Formula: C15H11N3O Molecular Weight (g/mol): 249.273 MDL Number: MFCD00004071 InChI Key: RAXUMGMWXZYADR-OBGWFSINSA-N Synonym: 1-2-pyridylazo-2-naphthol,unii-095b53y3xv,pan indicator,2-naphthalenol, 1-2-pyridinylazo,1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one,1-2-pyridylazo-2-hydroxynaphthalene,pan van,1-2-pyridyldiazenyl naphthalen-2-ol,1-pyridin-2-azo-2-naphthol,2-naphthalenol, 1-2-2-pyridinyl diazenyl PubChem CID: 5376264 IUPAC Name: (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one SMILES: C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3