Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN Indicator, 0.1% Solution in Ethanol, Spectrum™ Chemical
SDP

Catalog No. 18604956 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-956 500 mL Poly Bottle
18-604-955 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-604-956 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P575500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P-575, 85-85-8

Spectrum™ Chemical PAN Indicator, 0.1 Percent Solution in Ethanol

Chemical Identifiers

CAS 85-85-8 , 67-56-1
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-SDXDJHTJSA-N
IUPAC Name (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1

Specifications

CAS Max % 0.999, 0.001
Boiling Point 79°C/174.2°F
Packaging Poly Bottle
Quantity 500 mL
UN Number UN1170
Grade Reagent
Physical Form Liquid
Chemical Name or Material PAN Indicator, 0.1 Percent Solution in Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.