CAS RN 850140-72-6
IUPAC Name:
N-{4-[(3-chloro-4-fluorophenyl)amino]-7-(oxolan-3-yloxy)quinazolin-6-yl}-4-(dimethylamino)but-2-enamide
Synonyms:
afatinib
Molecular Weight (g/mol):
485.94
Molecular Formula:
C24H25ClFN5O3
InChi Key:
ULXXDDBFHOBEHA-UHFFFAOYNA-N
SMILES:
CN(C)CC=CC(=O)NC1=C(OC2CCOC2)C=C2N=CN=C(NC3=CC(Cl)=C(F)C=C3)C2=C1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Afatinib
Thermo Scientific Chemicals Afatinib
CAS: 850140-72-6 Molecular Formula: C24H25ClFN5O3