accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85017-82-9

IUPAC Name: lithium(1+) bromide
Synonyms: lithium bromide|lithium monobromide
Molecular Weight (g/mol): 86.84
Molecular Formula: BrLi
InChi Key: AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES: [Li+].[Br-]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)