Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bromide hydrate, Puratronic™, 99.995% (metals basis)

Catalog No. AA1073322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1073322 100 g
AA1073314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1073322 Supplier Thermo Scientific Chemicals Supplier No. 01073322
Add to Cart

CAS: 85017-82-9 | BrLi | 86.84 g/mol

Lithium bromide is used as a medium for air-conditioning, and a desiccant industrial drying system. It finds use as a sedative and hypnotic. Further it is employed in manufacturing pharmaceuticals, in alkylation processes, and as brazing and welding fluxes. In addition, lithium bromide-water solution is widely used in absorption refrigeration systems, absorption heat pumps and absorption heat transformers. l-Proline lithium bromide monohydrate (LPLBM) is a promising semi-organic nonlinear optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-82-9
Molecular Formula BrLi
Molecular Weight (g/mol) 86.84
MDL Number MFCD00149763
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
IUPAC Name lithium(1+) bromide
SMILES [Li+].[Br-]

Specifications

Melting Point 162°C to 167°C
Color White
Physical Form Crystalline Aggregates
Quantity 100 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water,alcohol and glycol.
Formula Weight 86.85 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lithium bromide hydrate
Hazard Category H302-H315-H319-H334
Precautionary Statement P261-P264b-P270-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
EINECSNumber 231-439-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.