accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850375-05-2

CH3NSO

CAS RN 850375-05-2

IUPAC Name: 3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
Synonyms: 3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
Molecular Weight (g/mol): 203.26
Molecular Formula: C11H9NOS
InChi Key: FQQPVEKQPGDYPD-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC(C=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results