Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Methyl-thiazol-4-yl)-benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC19404DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19404DE 5 g
CC19404CB 250 mg
CC19404DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC19404DE Supplier Thermo Scientific Chemicals Supplier No. CC19404DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850375-05-2
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.26
MDL Number MFCD02681946
InChI Key FQQPVEKQPGDYPD-UHFFFAOYSA-N
PubChem CID 7127782
IUPAC Name 3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
SMILES CC1=NC(=CS1)C1=CC(C=O)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 3-2-methyl-thiazol-4-yl-benzaldehyde, 3-2-methyl-1,3-thiazol-4-yl benzaldehyde, 3-2-methylthiazol-4-yl benzaldehyde, 4-3-formylphenyl-2-methyl-1,3-thiazole, 3-2-methyl-4-thiazolyl benzaldehyde, benzaldehyde,3-2-methyl-4-thiazolyl, benzaldehyde, 3-2-methyl-4-thiazolyl
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-thiazol-4-yl)-benzaldehyde
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water fo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.